Skip to Content
Merck
All Photos(1)

Key Documents

342000

Sigma-Aldrich

Caspase Inhibitor, Negative Control

The Caspase Inhibitor, Negative Control, also referenced under CAS 105637-38-5, controls the biological activity of Caspase. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

Caspase Inhibitor, Negative Control, Z-FA-FMK, Cathepsin B, Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23FN2O4
CAS Number:
Molecular Weight:
386.42
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

off-white

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C21H23FN2O4/c1-15(19(25)13-22)23-20(26)18(12-16-8-4-2-5-9-16)24-21(27)28-14-17-10-6-3-7-11-17/h2-11,15,18H,12-14H2,1H3,(H,23,26)(H,24,27)

InChI key

ASXVEBPEZMSPHB-UHFFFAOYSA-N

General description

A cell-permeable, irreversible cathepsin B inhibitor. Blocks LPS-induced production of IL-1α and IL-1β. Suitable as a negative control for caspase-1 and caspase-2 inhibitors because it lacks an aspartic acid residue at the P1 position.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Cathepsin-B
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Sequence

Z-Phe-Ala-FMK

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Schotte, P., et al. 2001. J. Biol. Chem.276, 21153.
Guo, H., et al. 1998. Exp. Cell Res.245, 57.
Rosenthal, P.J., et al. 1993. J. Clin. Invest.91, 1057.
Ahmed, N.K., et al. 1992. Biochem. Pharmacol.44, 1201.
Rauber, P., et al. 1986. Biochem. J. 239, 633.
Shaw, E., et al. 1986. Biomed. Biochim. Acta 45, 1397.
Rasnick, D. 1985. Anal. Biochem. 149, 461.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service