Skip to Content
Merck
All Photos(1)

Key Documents

317642

Sigma-Aldrich

Dipeptidylpeptidase IV Inhibitor IV, K579

The Dipeptidylpeptidase IV Inhibitor IV, K579, also referenced under CAS 440100-64-1, controls the biological activity of Dipeptidylpeptidase IV. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

Dipeptidylpeptidase IV Inhibitor IV, K579, (S)-1-(4-Methyl-1-(2-pyrimidinyl)-4-piperidylamino)acetyl-2-pyrrolidinecarbonitrile, DPP IV Inhibitor IV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H24N6O
CAS Number:
Molecular Weight:
328.41
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white to light brown

solubility

1 M HCl: 16 mg/mL
DMSO: 32 mg/mL
ethanol: 6.5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

N3([C@@H](CCC3)C#N)C(=O)CNC1(CCN(CC1)c2ncccn2)C

InChI

1S/C17H24N6O/c1-17(21-13-15(24)23-9-2-4-14(23)12-18)5-10-22(11-6-17)16-19-7-3-8-20-16/h3,7-8,14,21H,2,4-6,9-11,13H2,1H3/t14-/m0/s1

InChI key

JERPHRNGJBSFAP-AWEZNQCLSA-N

General description

A cyanopyrrolidine compound that acts as a potent DPP IV inhibitor (IC50 = 3, 5, 8, and 8 nM in inhibiting rat, human, monkey, and canine DDP IV, respectively). Oral administration of K579 in rats in vivo has been shown to reduce blood DPP IV activity and augment GLP-1 and insulin response after glucose intake and significantly reduce plasma glucose concentration.
Please note: 328.4 is the anhydrous molecular weight; please refer to the vial label for the lot-specific hydration and molecular weight.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Please note: 328.4 is the anhydrous molecular weight; please refer to the vial label for the lot-specific hydration and molecular weight.
Takasaki, K., et al. 2004. Eur. J. Pharmacol.505, 237.
Takasaki, K., et al. 2004. Eur. J. Pharmacol.486, 335.
Takasaki, K., et al. 2004. J. Pharmacol. Sci.95, 291.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service