Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
Product Name
NovaSeptum® Cell Culture Sampling Unit; Accurate Volume Application, 20mL
material
syringe (polycarbonate, medical grade, platinum-cured silicone, medical silicone fluid)
Quality Segment
product line
NovaSeptum®
manufacturer/tradename
NovaSeptum®
storage temp.
10-30°C
Application
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| product line NovaSeptum® | product line NovaSeptum® | product line NovaSeptum® | product line - |
| material syringe (polycarbonate, medical grade, platinum-cured silicone, medical silicone fluid) | material 316 stainless steel cannula (ASTM), polyester body, septum (Medical grade platinum-cured silicone) | material 316 stainless steel cannula (ASTM), polyester body, septum (Medical grade platinum-cured silicone) | material polycarbonate syringe (medical grade), polyethylene bag (drainage), silicone syringe (platinum-cured), silicone tubing (medical grade, platinum-cured), syringe (with medical silicone fluid) |
| manufacturer/tradename NovaSeptum® | manufacturer/tradename NovaSeptum® | manufacturer/tradename NovaSeptum® | manufacturer/tradename NovaSeptum® |
| Quality Level 400 | Quality Level 400 | Quality Level 400 | Quality Level 400 |
| storage temp. 10-30°C | storage temp. 10-30°C | storage temp. 10-30°C | storage temp. 10-30°C |
Still not finding the right product?
Explore all of our products under NovaSeptum® Cell Culture Sampling Unit; Accurate Volume Application
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


