219492
Cdk4/6 Inhibitor IV
The Cdk4/6 Inhibitor IV, also referenced under CAS 359886-84-3, controls the biological activity of Cdk4/6. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Synonym(s):
Cdk4/6 Inhibitor IV, trans-4-((6-(ethylamino)-2-((1-(phenylmethyl)-1H-indol-5-yl)amino)-4-pyrimidinyl)amino)-cyclohexanol, CINK4
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
Assay
≥95% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
pale amber
solubility
DMSO: 10 mg/mL
shipped in
ambient
storage temp.
2-8°C
General description
A cell-permeable triaminopyrimidine compound that acts as a reversible and ATP-competitive inhibitor against Cyclin D1-complexed, but not Cyclin D2-complexed, Cdk4 and Cdk6 (IC50 = 1.5 & 5.6 µM, respectively) with much reduced activity against Cdk5/p35 (IC50 = 25 µM) and little or no activity towards Cdk1/CycB,
Cdk2/CycA, Cdk2/CycE, v-abl, c-met, IGF-1R, or IR (IC50 >10 µM). CINK4 (at 5 & 10 µM) effectively inhibits cellular Cdk4 substrate pRb phosphorylation and prevents serum-stimulated G1/S transition of serum starved pRb-positive, but not pRb-negative, cultures. CINK4 is shown to exert a ~46% suppression of human colon carcinoma HCT116-derived tumor growth in mice in vivo (30 mg/kg; twice a day i.p.).
Cdk2/CycA, Cdk2/CycE, v-abl, c-met, IGF-1R, or IR (IC50 >10 µM). CINK4 (at 5 & 10 µM) effectively inhibits cellular Cdk4 substrate pRb phosphorylation and prevents serum-stimulated G1/S transition of serum starved pRb-positive, but not pRb-negative, cultures. CINK4 is shown to exert a ~46% suppression of human colon carcinoma HCT116-derived tumor growth in mice in vivo (30 mg/kg; twice a day i.p.).
A cell-permeable triaminopyrimidine compound that acts as a reversible and ATP-competitive inhibitor against Cyclin D1-complexed, but not Cyclin D2-complexed, Cdk4 and Cdk6 (IC50 = 1.5 and 5.6 µM, respectively) with much reduced activity against Cdk5/p35 (IC50 = 25 µM) and little or no activity towards Cdk1/CycB, Cdk2/CycA, Cdk2/CycE, v-abl, c-met, IGF-1R, or IR (IC50 >10 µM). CINK4 (at 5 and 10 µM) effectively inhibits cellular Cdk4 substrate pRb phosphorylation and prevents serum-stimulated G1/S transition of serum starved pRb-positive, but not pRb-negative, cultures. CINK4 is shown to exert a ~46% suppression of human colon carcinoma HCT116-derived tumor growth in mice in vivo (30 mg/kg; twice a day, i.p.).
Packaging
Packaged under inert gas
Warning
Toxicity: Standard Handling (A)
Other Notes
Soni, R., et al. 2001. J. Natl. Cancer Inst.93, 436.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Autophagy, 18(8), 1785-1800 (2021-11-17)
Modulation of the host cell cycle has emerged as a common theme among the pathways regulated by bacterial pathogens, arguably to promote host cell colonization. However, in most cases the exact benefit ensuing from such interference to the infection process
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service