Skip to Content
Merck
All Photos(1)

Key Documents

219445

Sigma-Aldrich

Cdk2 Inhibitor II

The Cdk2 Inhibitor II, also referenced under CAS 222035-13-4, controls the biological activity of Cdk2. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Cdk2 Inhibitor II, Compound 3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11BrN4O3S
CAS Number:
Molecular Weight:
395.23
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (sum of two isomers, HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable, reversible, ATP-competitive, potent and selective inhibitor of Cdk2 (IC50 = 60 nM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
CDK2
Product competes with ATP.
Reversible: yes
Target IC50: 60 nM against Cdk2

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Davis, S.T., et al. 2001. Science291, 134.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The TRESLIN-MTBP complex couples completion of DNA replication with S/G2 transition.
Zonderland, et al.
Molecular Cell, 82, 3350-3365 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service