Skip to Content
Merck
All Photos(1)

Documents

217594

Sigma-Aldrich

Anti-CD44 Rat mAb (A020)

liquid, clone A020, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

A020, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

canine, bovine, human, rabbit, porcine, mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG2b

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

bovine ... Cd44(281057)
dog ... Cd44(403939)
human ... CD44(960)
mouse ... Cd44(12505)
pig ... Cd44(100525150)

General description

Anti-CD44, rat monoclonal, clone A020, recognizes the ~85-95 kDa CD44 protein (gp90) in human and GP85 in mouse. It is validated for ELISA, IF, IP, RIA, WB, and IHC.
Rat monoclonal antibody purified by ammonium sulfate precipitation and DEAE chromatography. Recognizes the ~85-95 kDa CD44 protein.
Recognizes the ~85-95 kDa CD44 protein (gp90) in human and GP85 or Pgp-1 in mouse.

Immunogen

Human
purified human lymphocyte CD44

Application

ELISA (1:2000)

Flow Cytometry (1:1000)

Immunoblotting (1:1000)

Immunofluorescence (1:200)

Immunohistochemistry (1:250)

Immunoprecipitation (1:200)

Neutralization Studies (see comments and application references)

Radioimmunoassay (1:5000)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 150 mM NaCl, 10 mM sodium phosphate, pH 7.5.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Iida, N., and Bourguignon, L.Y.W. 1995. J. Cell Physiol.162, 121.
Bourguignon, L.Y.W., et al. 1993. J. Immunol.151, 6634.
The antibody specifically recognizes and reacts with a common determinant of the CD44 class of glycoproteins. This antibody has been reported to block the interaction between hyaluronan and CD44 (see application references). Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eun-Ju Chang et al.
Journal of cell science, 120(Pt 1), 166-176 (2006-12-14)
The differentiation of osteoclasts, cells specialized for bone resorption, is governed by two key factors, macrophage colony stimulating factor (M-CSF) and receptor activator of nuclear factor kappaB ligand (RANKL). The extracellular matrix (ECM) is an important factor influencing cell fate.
Daniel S Puperi et al.
Biomacromolecules, 17(5), 1766-1775 (2016-04-28)
Advanced tissue engineered heart valves must be constructed from multiple materials to better mimic the heterogeneity found in the native valve. The trilayered structure of aortic valves provides the ability to open and close consistently over a full human lifetime
Shibnath Ghatak et al.
The Journal of biological chemistry, 280(10), 8875-8883 (2005-01-06)
Hyaluronan is enriched in many types of human cancers, and manipulations of hyaluronan expression or interactions have a major influence on tumor progression in animal models. Increased ErbB2 activity is characteristic of several cancers and is responsible for many aspects
Mengya Zhao et al.
Cell reports, 42(9), 113038-113038 (2023-08-25)
Chronic neurodegeneration and acute injuries lead to neuron losses via diverse processes. We compared retinal ganglion cell (RGC) responses between chronic glaucomatous conditions and the acute injury model. Among major RGC subclasses, αRGCs and intrinsically photosensitive RGCs (ipRGCs) preferentially survive
Dong Duan et al.
Asian Pacific journal of cancer prevention : APJCP, 13(9), 4255-4261 (2012-11-22)
The present study was conducted to evaluate radioimmunoimaging (RII) and in vivo distribution of mixed antibodies 99mTc-EGFR-mAb and 99mTc-CD44- mAb in nude mice bearing human lung adenocarcinoma xenografts. Single and mixed applications of the two radiolabeled monoclonal antibodies (mAbs) were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service