Skip to Content
Merck
All Photos(1)

Key Documents

208665

Sigma-Aldrich

Calmidazolium Chloride

A cell-permeable calmodulin antagonist.

Synonym(s):

Calmidazolium Chloride, Compound R 24571

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H23Cl7N2O
CAS Number:
Molecular Weight:
687.70
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 10 mg/mL
ethanol: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C31H24Cl6N2O.Cl/c32-23-6-1-20(2-7-23)31(21-3-8-24(33)9-4-21)39-14-13-38(19-39)17-30(27-12-11-26(35)16-29(27)37)40-18-22-5-10-25(34)15-28(22)36;/h1-16,30-31H,17-19H2;

InChI key

LBFGPQFQXRJXDI-UHFFFAOYSA-N

General description

A cell-permeable calmodulin antagonist. An analog of sepazonium that is at least 150 times more potent than Trifluoperazine (Cat. No. 642150) as an inhibitor of brain Ca2+-calmodulin-dependent phosphodiesterase (IC50 = 10 nM). Inhibits voltage-gated Ca2+ channels and stimulates the release of nitric oxide in neuroblastoma cells. Also, acts as a strong inhibitor of calcium-dependent Ca2+-ATPase in erythrocytes. Known to prolong cardiac refractory period in vivo.
A cell-permeable calmodulin antagonist. At least 150 times more potent than Trifluoperazine (Cat. No. 642150) as an inhibitor of brain calmodulin-dependent phosphodiesterase (IC50 = 10 nM). An inhibitor of voltage-gated Ca2+ channels. Stimulates the release of nitric oxide in neuroblastoma cells. Also acts as a strong non-competitive inhibitor of skeletal muscle sarcoplasmic reticulum Ca2+-ATPase (Ki = 60 nM). Known to prolong cardiac refractoriness in vivo.
Note: easily adheres to glass in aqueous solution.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
brain calmodulin-dependent phosphodiesterase
Product does not compete with ATP.
Reversible: no
Target IC50: 10 nM against brain calmodulin-dependent phosphodiesterase

Caution

readily adheres to glass in aqueous solution.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C) for long term storage or refrigerate (4°C) for short-term storage. DMSO stock solutions are stable for up to 2 months -20°C.

Other Notes

Hu, J., and El-Fakahany, E.E. 1993. NeuroReport. 4, 198.
Nakazawa, K., et al. 1993. Br. J. Pharmacol. 109, 137.
Yuasa, T., and Muto, S. 1992. Arch. Biochem. Biophys.296, 175.
Ichikawa, M., et al. 1991. J. Membrane Biol.120, 211.
Silver, P.J., et al. 1989. Cardiovasc. Drug Ther. 3, 675.
Anderson, K.W., et al. 1984. J. Biol. Chem.259, 11487.
Gietzen, L., et al. 1982. Biochem. J. 207, 541.
Van Belle, H. 1981. Cell Calcium2, 483.
Levin, R.N., and Weiss, B. 1979. J. Pharmacol. Exp. Therap.208, 454.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Corentin Léger et al.
BMC biology, 20(1), 176-176 (2022-08-10)
Calmodulin (CaM) is an evolutionarily conserved eukaryotic multifunctional protein that functions as the major sensor of intracellular calcium signaling. Its calcium-modulated function regulates the activity of numerous effector proteins involved in a variety of physiological processes in diverse organs, from
Dechen Fu et al.
Cell death & disease, 13(1), 63-63 (2022-01-20)
Colorectal cancer (CRC) is one of the most common and deadliest forms of cancer. Myeloid Cell Leukemia 1 (MCL1), a pro-survival member of the Bcl-2 protein family is associated with chemo-resistance in CRC. The ability of MCL1 to inhibit apoptosis
Roseleen Ekka et al.
PLoS pathogens, 20(6), e1012303-e1012303 (2024-06-17)
Chlamydia trachomatis is a clinically important bacterium that infects epithelial cells of the genitourinary and respiratory tracts and the eye. These differentiated cells are in a quiescent growth state and have a surface organelle called a primary cilium, but the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service