Skip to Content
Merck

Skip To

20-175

HDAC Assay Buffer, 5X

The protein encoded by this gene belongs to the class II histone deacetylase/acuc/apha family. It possesses histone deacetylase activity & represses transcription when tethered to a promoter.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
41116012
NACRES:
NA.84
eCl@ss:
32160405
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

form

liquid

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (histone deacetylase (HDAC))

NCBI accession no.

shipped in

wet ice

Application

For use in the Histone Deacetylase Assay (HDAC) Kit.
Research Category
All
The protein encoded by this gene belongs to the class II histone deacetylase/acuc/apha family. It possesses histone deacetylase activity & represses transcription when tethered to a promoter.

Physical form

5X HDAc Assay Buffer: 50mM Tris-HCl, pH 8.0, 750mM NaCl and 50% glycerol.

Preparation Note

1 year at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
20-14820-14520-108
technique(s)

activity assay: suitable (histone deacetylase (HDAC))

technique(s)

activity assay: suitable (histone acetyltransferase (HAT))

technique(s)

activity assay: suitable (kinase)

technique(s)

activity assay: suitable (kinase)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

form

liquid

form

liquid

form

liquid

form

liquid

manufacturer/tradename

Upstate®

manufacturer/tradename

Upstate®

manufacturer/tradename

Upstate®

manufacturer/tradename

Upstate®

UniProt accession no.

O15379, Q13547, Q92769, Q96DB2, Q9UBN7, Q9UQL6, P56524, Q969S8, Q9UKV0, Q9BY41, Q8WUI4

UniProt accession no.

-

UniProt accession no.

-

UniProt accession no.

-

NCBI accession no.

NM_004964.2

NCBI accession no.

-

NCBI accession no.

-

NCBI accession no.

-


Storage Class

10 - Combustible liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



H Inadome et al.
Bioscience, biotechnology, and biochemistry, 65(11), 2577-2580 (2002-01-17)
A previously uncharacterized yeast protein, YJL066c, was discovered in the membrane fraction although it has no hydrophobic stretch. The protein was partly solubilized by Triton X-100 in an oligomeric form, while it was insoluble in alkali or salt. By immunofluorescent



Global Trade Item Number

SKUGTIN
20-17504053252032769
20255-U04061833524657
Z547700-1EA04061832112817
MTCC0175-120EA04065272294935

Questions

Reviews

No rating value

Active Filters