Skip to Content
Merck
All Photos(1)

Documents

196440

Sigma-Aldrich

Batimastat

≥98% (HPLC), solid, Matrix metalloproteinase inhibitor, Calbiochem

Synonym(s):

Batimastat, (4-N-Hydroxyamino)-2 R-isobutyl-3 S-(thienylthiomethyl)succinyl)-L-phenylalanine-N-methylamide, BB-94, (4-N-Hydroxyamino)-2R-isobutyl-3S-(thienylthiomethyl)succinyl)-L-phenylalanine-N-methylamide, BB-94

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H31N3O4S2
CAS Number:
Molecular Weight:
477.64
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

product name

Batimastat, Batimastat primarily used in Inhibition.

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C23H31N3O4S2/c1-15(2)12-17(18(22(28)26-30)14-32-20-10-7-11-31-20)21(27)25-19(23(29)24-3)13-16-8-5-4-6-9-16/h4-11,15,17-19,30H,12-14H2,1-3H3,(H,24,29)(H,25,27)(H,26,28)/t17-,18+,19+/m1/s1

InChI key

XFILPEOLDIKJHX-QYZOEREBSA-N

General description

A Marimastat (Cat. No. 444289) type of peptidyl hydroxamate-based inhibitor that potently inhibits a broad-spectrum of metalloproteinases, including MMP-1, MMP-2, MMP-3/stromelysin, MMP-7/matrilysin, MMP-9, ΔMT1 (MMP-14 without TM domain), ADAM8, and ADAM17/TACE (IC50 = 3, 4, 20, 6, 4, 2.08, 51.3, and 19 nM, respectively), by targeting both the substrate binding site and the active-site Zn2+, while being much less potent toward ACE (Angiotensin Converting Enzyme) or α-secretase (IC50 = 1.6 and 3.3 µM, respectively). Batimastat is widely used in studying the involvement of MMPs in cancinogenesis and non-cancer pathological processes both in cultures in vitro and in animals in vivo. Also available as a 25 mM solution in DMSO (Cat. No. 508408).
A Marimastat (Cat. No. 444289) type of peptidyl hydroxamate-based inhibitor that potently inhibits a broad-spectrum of metalloproteinases, including MMP-1, MMP-2, MMP-3/stromelysin, MMP-7/matrilysin, MMP-9, ΔMT1 (MMP-14 without TM domain), ADAM8, and ADAM17/TACE (IC50 = 3, 4, 20, 6, 4, 2.08, 51.3, and 19 nM, respectively), by targeting both the substrate binding site and the active-site Zn2+, while being much less potent toward ACE (Angiotensin Converting Enzyme) or α-secretase (IC50 = 1.6 and 3.3 µM, respectively). Batimastat is widely used in studying the involvement of MMPs in cancinogenesis and non-cancer pathological processes both in cultures in vitro and in animals in vivo.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Schlomann, U., et al. 2002. J. Biol. Chem.277, 48210.
Whittaker, M., et al. 1999. Chem. Rev.99, 2735.
Parvathy, S., et al. 1998. Biochemistry37, 1680.
Parvathy, S., et al. 1998. FEBS Lett.431, 63.
Yamamoto, M., et al. 1998. J. Med. Chem.41, 1209.
Moss, M.L., et al. 1997. Nature385, 733.
Eccles, S.A., et al. 1996. Cancer Res.56, 2815.
Brown, P.D. 1995. Advan. Enzyme Regul.35, 293.
Wang, X., et al. 1994. Cancer Res.54, 4726.
Davies, B., et al. 1993. Cancer Res.53, 2087.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abdulbasit Amin et al.
Molecular metabolism, 73, 101731-101731 (2023-05-01)
The metalloprotease ADAM17 (also called TACE) plays fundamental roles in homeostasis by shedding key signaling molecules from the cell surface. Although its importance for the immune system and epithelial tissues is well-documented, little is known about the role of ADAM17
Ivonne Melano et al.
Microbiology spectrum, 11(5), e0385422-e0385422 (2023-09-15)
Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), the etiological agent of the Coronavirus disease-19 (COVID-19) pandemic, utilizes angiotensin-converting enzyme 2 (ACE2) as a receptor for virus infection. However, the expression pattern of ACE2 does not coincide with the tissue tropism
Jana Koch et al.
Cancers, 14(15) (2022-08-13)
Matrix metalloproteinases (MMPs) play a crucial role in tumour initiation, progression, and metastasis, including peritoneal carcinosis (PC) formation. MMPs serve as biomarkers for tumour progression in colorectal cancer (CRC), and MMP overexpression is associated with advanced-stage metastasis and poor survival.
Marina Badenes et al.
Life science alliance, 6(4) (2023-02-01)
The metalloprotease ADAM17 is a sheddase of key molecules, including TNF and epidermal growth factor receptor ligands. ADAM17 exists within an assemblage, the "sheddase complex," containing a rhomboid pseudoprotease (iRhom1 or iRhom2). iRhoms control multiple aspects of ADAM17 biology. The
Eva Maria Wenzel et al.
Nature communications, 15(1), 1277-1277 (2024-02-11)
Overexpression of the transmembrane matrix metalloproteinase MT1-MMP/MMP14 promotes cancer cell invasion. Here we show that MT1-MMP-positive cancer cells turn MT1-MMP-negative cells invasive by transferring a soluble catalytic ectodomain of MT1-MMP. Surprisingly, this effect depends on the presence of TKS4 and

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service