Skip to Content
Merck
All Photos(1)

Key Documents

1.94939

Sigma-Aldrich

Hydrazine hydrate

80%, for analysis EMPARTA®

Synonym(s):

Hydrazine hydroxide, Hydrazinium hydroxide

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352306
Pricing and availability is not currently available.

Product Name

Hydrazine hydrate 80%, for analysis EMPARTA®

Quality Level

storage temp.

room temp

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service