Skip to Content
Merck
All Photos(1)

Documents

1.88075

Supelco

Hose connection for Titripac

hose L 80 cm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116105
NACRES:
NA.21

product line

Titripur®

technique(s)

titration: suitable

hose L

80 cm

storage temp.

15-25°C

General description

(Length 80 cm) inclusive adapter Titripur®

Legal Information

Titripur is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service