Skip to Content
Merck
All Photos(1)

Key Documents

182707

Sigma-Aldrich

GNF351

≥98% (HPLC), solid, AhR antagonist, Calbiochem®

Synonym(s):

AhR Antagonist III, GNF351, N-(2-(3H-Indol-3-yl)ethyl)-9-isopropyl-2-(5-methyl-3-pyridyl)-7H-purin-6-amine, N-(2-(1H-Indol-3-yl)ethyl)-9-isopropyl-2-(5-methylpyridin-3-yl)-9H-purin-6-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H25N7
CAS Number:
Molecular Weight:
411.50
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

AhR Antagonist III, GNF351, AhR Antagonist III, GNF351, is a cell-permeable, high-affinity aryl hydrocarbon receptor (AhR) antagonist (IC50 = 62 nM in mouse liver cytosol expressing humanized AhR).

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

shipped in

wet ice

storage temp.

2-8°C

SMILES string

[n]1(c2nc(nc(c2nc1)NCCc4c5c([nH]c4)cccc5)c3cncc(c3)C)C(C)C

InChI

1S/C24H25N7/c1-15(2)31-14-28-21-23(26-9-8-17-13-27-20-7-5-4-6-19(17)20)29-22(30-24(21)31)18-10-16(3)11-25-12-18/h4-7,10-15,27H,8-9H2,1-3H3,(H,26,29,30)

InChI key

ABXIUYMKZDZUDC-UHFFFAOYSA-N

General description

A cell-permeable purine compound that acts as a high-affinity aryl hydrocarbon receptor (AhR) antagonist (IC50 = 62 nM in mouse liver cytosol expressing humanized AhR) and lacks any agonist activity even at higher doses. Reportedly binds with high affinity to the ligand-binding pocket of AhR and blocks the binding of an array of exogenous and endogenous ligands. Shown to non-covalently interact with Ser317, His291, and Ser365 in human and with Ser311, His285, and Ser359 in mouse AhR. Represses AhR transcriptional activity via the dioxin response element (DRE)-dependent and independent mediated responses in human and mouse cells (IC50 = 8.5 nM in HepG2 40/6 cells).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Smith, K.J., et al. 2011. J. Pharmacol. Exp. Ther. 338, 318.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service