Skip to Content
Merck
All Photos(2)

Key Documents

182541

Sigma-Aldrich

Aromatase Inhibitor II, Letrozole

The Aromatase Inhibitor II, Letrozole, also referenced under CAS 112809-51-5, controls the biological activity of Aromatase.

Synonym(s):

Aromatase Inhibitor II, Letrozole, 4,4ʹ-(1H-1,2,4-Triazol-1-ylmethylene)dibenzonitrile, 1-( bis-(4-Cyanophenyl)methyl)-1,2,4-triazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H11N5
CAS Number:
Molecular Weight:
285.30
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥99% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

ethanol: <5 mg/mL (partially soluble)
DMSO: 50 mg/mL, clear, colorless

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C17H11N5/c18-9-13-1-5-15(6-2-13)17(22-12-20-11-21-22)16-7-3-14(10-19)4-8-16/h1-8,11-12,17H

InChI key

HPJKCIUCZWXJDR-UHFFFAOYSA-N

General description

An orally bioavailable non-steroidal triazole compound that acts as a potent and selective inhibitor of aromatase (IC50 = 140, 350 and 450 pM in Dex-treated fibroblasts, MCF-7Ca, and JEG-3 cells, respectively). Competitively, but reversibly binds to the heme iron of aromatase. Does not affect the synthesis of adrenal mineralocorticoid or glucocorticoids. Reported to be highly effective as an anti-estrogenic and anti-proliferative agent in estrogen-rich tumors. Also delays the fusing of the growth plates in adolescents and enhances the effectiveness of growth hormone. Shown to synergistically induce apoptosis and cause tumor regression when combined with IGF-1R inhibitors.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Hou, X., et al. 2011. Cancer Res.71, 7597.
Lisztwan, J., et al. 2008. Breast Cancer Res.10, R56.
Miller, W.R., et al. 2002. Cancer Invest.20 (Suppl 2), 15.
Long, B.J., et al. 1998. J. Steroid Biochem. Molec. Biol.67, 293.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service