Skip to Content
Merck
All Photos(1)

Key Documents

1.71204

Supelco

PC cable for thermoreactors

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41102404

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® TR 420
for use with Spectroquant® TR 620

storage temp.

2-30°C

General description

TR 420, TR 620 Spectroquant®

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service