1.50179
LiChroCART® for customized packing HPLC Columns
L × I.D. 250 mm × 10 mm; LiChroCART® HPLC cartridge, manuCART™ holder required for use; please select HPLC sorbent for Column filling
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
product name
LiChroCART® 250-10, suitable for HPLC
parameter
250 bar pressure
Quality Level
technique(s)
HPLC: suitable (Semi Prep HPLC)
column L × I.D.
25 cm × 10 mm
storage temp.
no temp limit
Related Categories
General description
HPLC cartridge, customized packing, please specify packing material
Legal Information
LICHROCART is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service