Skip to Content
Merck
All Photos(1)

Key Documents

14-791-M

Sigma-Aldrich

PI 3-Kinase (p110α(E542K)/p85α) murine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf21 cells

Quality Level

description

protein target PI3Kα/p85α

purified by

(Ni2+/NTA-agarose)

UniProt accession no.

Gene Information

mouse ... PIK3CA(18706)

General description

Complex of N-terminal 6His-tagged recombinant murine p110α full length, containing the mutation E542K, and untagged, recombinant, murine p85α full length.
The E542K substitution is a somatic mutation in p110α that has been associated with tumours of the colon and brain. Combined in vitro and in vivo studies have shown that this mutation confers higher lipid kinase activity than wild type, and is able to induce oncogenic transformation. (Kang S. et al., PNAS, (2005);102: 802-807 and Zhao J.J. et al., PNAS, (2005);102:18443-18448).

Packaging

Also available in 250μg size --call for pricing and availability and reference catalog number 14-791M when ordering the 250μg size.
Also available in 10 μg size (cat#14-791) and in bulk (cat# 14-791-K).

Physical properties

p110α(E542K) MW = 129 kDa, p85α MW = 84 kDa.

Physical form

50 mM Tris/HCl pH7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 0.2 mM PMSF, 1 mM benzamidine, 0.1% 2-mercaptoethanol. Frozen solution.

Storage and Stability

Store at -70°C from date of shipment. For maximum recovery of product, centrifuge original vial prior to removing the cap.

Analysis Note

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service