Recommended Products
material
stainless steel
Quality Level
feature
autoclavable
L × W × H
19.6 cm × 9.9 cm × 10.4 cm
suitability
suitable for (to be used with the ICR swab)
application(s)
pharmaceutical
surface monitoring
compatibility
for use with ICR Swab
shipped in
ambient
storage temp.
no temp limit
Related Categories
General description
Application
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
The ICR-Swab is a swab system for environmental monitoring of dry surfaces, such as filling needles, equipment recesses, uneven surfaces and tubing in clean rooms and isolators.
Contact Plates and Swabs for Personnel and Surface Monitoring in Cleanrooms and Isolators
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service