1.46439
Tryptic Soy Broth Ready-to-use Bottles
Histidin 0.05 %, Tween® 1.5 %, Lecithin 0.1 %, bottle capacity 250 mL, bottle filling volume 180 mL, closure type, blue screw cap and 3 loci, box of 6 or 12 bottles
Synonym(s):
CASO, Casein Soybean Digest Broth with 0.1% Lecithin, 1.5% Tween, 0.05% Histidin, SCDB, TSB with neutralizer, Tryptic Soy Broth w. LTH
About This Item
Recommended Products
sterility
sterile; autoclaved
Quality Level
form
liquid
feature
closure type blue screw cap and 3 loci
ready-to-use
composition
Histidin, 0.05%
Lecithin, 0.1%
Tween®, 1.5%
packaging
box of 6 or 12 bottles
bottle capacity
250 mL
bottle filling volume
180 mL
pH
7.3
application(s)
cosmetics
food and beverages
pharmaceutical
sterility testing
suitability
nonselective for
Application
Packaging
Storage and Stability
Other Notes
Legal Information
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service