Skip to Content
Merck
All Photos(2)

Documents

1.46376

Millipore

Tryptic Soy Broth Ready-to-use Bottles

with Tween® 80 0.1 %, bottle capacity 1000 mL, bottle filling volume 1000 mL, closure type, blue screw cap with 3 loci, box of 6 bottles

Synonym(s):

CASO, Casein Soybean Digest Broth with 0.1% Polysorbate, SCDB, TSB, Tryptic Soy Broth with 0.1% Tween, Tryptic Soy Broth with 0.1% Tween® 80

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106214

Agency

EP 2.6.1
JP 4.06
USP 71
71

Quality Level

sterility

sterile; autoclaved

form

liquid

feature

closure type blue screw cap with 3 loci
ready-to-use

composition

with Tween® 80, 0.1%

packaging

box of 6 bottles

bottle capacity

1000 mL

bottle filling volume

1000 mL

pH

7.3±0.2

application(s)

cosmetics
food and beverages
pharmaceutical
sterility testing

storage temp.

2-25°C

suitability

nonselective for

Looking for similar products? Visit Product Comparison Guide

General description

Tryptic Soy Broth with 0.1% Tween® 80 is a complex nutrient rich inhibitor free microbial media designed in compliance with European (EP), Japanese (JP) and United States Pharmacopeia (USP).
The medium is supplemented with peptones from casein and soymeal to provide an enhanced carbon source, which promotes the growth of the microbes present in the test sample.
Polysorbate 80 (Tween®) acts as a neutralizer to inactivate potential residues of disinfectants containing active agents such as Alcohol (70% ethanol or isopropyl alcohol), Aldehyde, Dichloroisocyanurate, Glucoprotamine, Peracetic acid, Phenols, Biguanides and quaternary ammonium compounds.

Application

Tryptic Soy Broth with 0.1% Tween® 80 is used for the isolation and cultivation of fastidious bacteria, yeast. TSB is suitable for sterility testing of substances, preparations, and products from pharmaceutical and food industry.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service