Skip to Content
Merck
All Photos(1)

Key Documents

1.46154

Millipore

Plate Count Agar Contact

55 mm Contact Plates for the determination of total count of aerobic bacteria, Room temperature storage

Synonym(s):

Tryptone Glucose Yeast Agar, Casein-Peptone Dextrose Yeast Agar, Standard Methods Agar, PCA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74
Pricing and availability is not currently available.

material

polystyrene plate

Quality Level

Agency

ISO

sterility

sterile; aseptically filled

form

plate (ready-to-use)

feature

2D bar code
ready-to-use
ICR Contact Plate for Room temperature with non-lockable lid

packaging

single packed of 20 plates

plate diam.

55 mm

application(s)

microbiology
pharmaceutical
surface monitoring

compatibility

for use with Contact Plate

storage temp.

15-25°C

suitability

aerobic bacteria (total count)

General description

Plate Count Agar Contact - RT is designed for the determination of the total aerobic bacteria on dry, sanitized surfaces and personnel in the non-critical area.
Plate Count Agar plates are used for the enumeration of the total viable count. The pH in the range of 6.8-7.2 is optimum for the growth of bacteria. The media is relatively rich in its nutrients and does not contain any inhibitors or indicators. The enzymatic digest of casein serves as a nitrogen source with high levels of free amino acids followed by yeast extract, which is a rich source of vitamins. The carbon source for the growth of bacteria is served by glucose, thereby providing efficient growth of bacteria.

Application

The Plate Count Agar is suitable for the determination of the total count of aerobic bacteria on dry, sanitized surfaces and personnel in the non-critical area.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Contact Plates and Swabs for Personnel and Surface Monitoring in Cleanrooms and Isolators

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service