Skip to Content
Merck
All Photos(1)

Key Documents

1.46139

Millipore

Fluid Thioglycollate Medium

tube capacity 17 mL, tube filling volume 10 mL, closure type, Red scew cap with 2 loci, pack of 100 tubes

Synonym(s):

Fluid Thioglycollate Medium, Thioglycollate Broth, FTM, Thioglycollate Medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106225
NACRES:
NA.74
Pricing and availability is not currently available.

Agency

EP 2.6.1
JP 4.06
USP 71

Quality Level

sterility

sterile; autoclaved

form

liquid (ready-to-use)

shelf life

6 mo.

feature

closure type Red scew cap with 2 loci
ready-to-use

packaging

pack of 100 tubes

capacity

17 mL

tube capacity

17 mL

tube filling volume

10 mL

pH

7.1

application(s)

cosmetics
pharmaceutical
sterility testing

storage temp.

2-25°C

suitability

nonselective for

Looking for similar products? Visit Product Comparison Guide

General description

Fluid Thioglycollate Medium is a universal complex medium. Thioglycollate and L-Cystine reduce the redox potential of the culture medium and create an anaerobic atmosphere, along with inactivating mercury and other heavy metal compounds. The presence of agar reduces a rapid diffusion of oxygen through the medium but may lead to slight turbidity in larger volumes (filled bottles). Resazurin indicates the reduction potential of the medium. An increased concentration of oxygen is indicated by a color change to pink.

Application

Fluid Thioglycollate Medium is used for the isolation and cultivation of fastidious anaerobic and aerobic microorganisms. It is suitable for sterility control of pharmaceutical products.

Features and Benefits

Our ready to use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopoeia requirements. Sterility testing media and rinse solutions are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility and growth promotion tests. Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, therefore improving accuracy and significantly reducing the risk of incorrect interpretation and false results.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Sterility testing media options facilitate direct inoculation for non-filterable samples, including soybean casein digest and Fluid Thioglycollate medium.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service