Skip to Content
Merck
All Photos(2)

Key Documents

1.46003

Millipore

Sabouraud Dextrose Agar

Chloramphenicol, EP, lid, ICR plus: lockable settle plate for long incubation, plate diam. 90 mm, sterile; aseptically filled, single packed of 20 and 120 plates, suitable for surface monitoring

Synonym(s):

SDA Settle Plate, SDA with Chloramphenicol

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74
Pricing and availability is not currently available.

material

polystyrene plate

Quality Level

Agency

EP
USP

sterility

sterile; aseptically filled

form

plate (ready-to-use)

feature

lid ICR plus: lockable settle plate for long incubation
ready-to-use

composition

Chloramphenicol

packaging

single packed of 20 and 120 plates

plate diam.

90 mm

pH

5.6 ( in H2O)

application(s)

air monitoring
cosmetics
food and beverages
pharmaceutical

surface monitoring

compatibility

for use with MAS-100®

storage temp.

15-25°C

suitability

molds
yeasts

Looking for similar products? Visit Product Comparison Guide

General description

Sabouraud Dextrose Agar + Chloramphenicol in 90 mm settle plate with 30 mL filling volume for long incubation is designed for the detection of yeasts and molds for air and personnel monitoring in less critical environments.
Sabouraud Dextrose Medium contains digests of animal tissues (peptones), which provide a nutritious source of amino acids and nitrogenous compounds for the growth of fungi and yeasts. Dextrose is the energy and carbon source. The low pH favors fungal growth and the addition of chloramphenicol inhibits the growth of the accompanying bacterial flora.

Application

Sabouraud Dextrose Agar + Chloramphenicol settle plate is designed for the detection of the total count of yeasts and molds in air and personnel of less critical environments.

Legal Information

MAS-100 is a registered trademark of MBV AG

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service