Skip to Content
Merck
All Photos(1)

Documents

14-558-M

Sigma-Aldrich

PI3 Kinase (p120γ), 20 µg

For use in Kinase Assays.

Synonym(s):

Active Kinase Protein, PI3 Kinase p120γ

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
eCl@ss:
32160405
NACRES:
NA.26

biological source

human

Quality Level

mol wt

Mw 130 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

Gene Information

human ... PIK3CG(5294)

General description

N-terminal His6-tagged recombinant full-length human PI 3-kinase (p120γ).

Packaging

Also available in 250μg size as catalog # 14-558M - Call for pricing and availability.

Quality

Routinely evaluated in the PIProfiler.

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nonradioactive methods for the assay of phosphoinositide 3-kinases and phosphoinositide phosphatases and selective detection of signaling lipids in cell and tissue extracts.
Gray, Alexander, et al.
Analytical biochemistry, 313, 234-245 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service