Skip to Content
Merck

1.37171

SAFC

Polysorbate 80

high purity, EMPROVE® EXPERT, Ph. Eur., ChP, JP, NF

Pharma Manufacturing

Synonym(s):

TWEEN® 80, POE (20) sorbitan monooleate, Polyethylene glycol sorbitan monooleate, Polyoxyethylenesorbitan monooleate, Polysorbate 80

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352207
EC Index Number:
500-019-9
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

GMP
Halal
Kosher

Agency

ChP
JP
NF
Ph. Eur.

description

GMO free
low in endotoxins

product line

EMPROVE® EXPERT

form

liquid

potency

42200 mg/kg LD50, oral

mol wt

1310 g/mol

pH

5-7 (20 °C, 50 g/L)

bp

>100 °C

density

1.07 g/cm3 at 25 °C

application(s)

liquid formulation
parenterals
pharmaceutical
solubility enhancement

foreign activity

Aflatoxin free

storage temp.

15-25°C

SMILES string

CCCCCCCC/C=C/CCCCCCCC(=O)OCCOCC(C1C(CC(O1)OCCO)OCCO)OCCO

InChI

1S/C32H60O10/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-30(36)40-25-24-37-27-29(39-22-19-34)32-28(38-21-18-33)26-31(42-32)41-23-20-35/h9-10,28-29,31-35H,2-8,11-27H2,1H3/b10-9+

InChI key

RGPBUVUVZKQNHD-MDZDMXLPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Polysorbate 80 is used to stabilize biomolecule formulations and as solubilizer, emulsifier and wetting agent in small molecule formulations. Polysorbates are known to degrade via autoxidation and hydrolysis. Degradation products can have a negative impact on the stability of the protein as well as the small molecule API. To reduce this risk, the polysorbate high-purity grade contains a low amount of impurities, especially peroxide.

Intended for high-risk applications, this Emprove® Expert grade contains bioburden and endotoxin limits.

Packaging

1371710100 / 100ml / glass bottle
1371711000 / 1L / glass bottle

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany
TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This article outlines critical factors in long-term stability of proteins, and how different stabilizers protect again mechanical and thermal stress.

Medicine for children poses unique formulation challenges compared to adults. Consider developmental physiology and age specifics when designing pharmaceuticals. Quality issues can severely impact patient safety. Therefore, excipient quality, supplier selection, and supply chain security are crucial, particularly for pediatric formulations.

Related Content

Learn more on mAb subcutaneous or intravenous formulation, and the relevant associated products, including excipients.

Developing formulations specifically for infants and children is increasingly important. To help you master these challenges effectively, we offer an extensive portfolio of well-established high-quality excipients and APIs for pediatric pharmaceutical formulations that are proven in practice.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service