Skip to Content
Merck
All Photos(1)

Key Documents

1.28210

SAFC

Zinc sulfate heptahydrate

FCC, GB

Pharma Manufacturing

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnSO4 · 7H2O
CAS Number:
Molecular Weight:
287.56
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-793-3
Pricing and availability is not currently available.

grade

GMO free
Halal
Kosher

Quality Level

Agency

GB

reg. compliance

FCC

form

powder

application(s)

food and beverages

storage temp.

2-30°C

SMILES string

O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O

InChI

1S/H2O4S.7H2O.Zn/c1-5(2,3)4;;;;;;;;/h(H2,1,2,3,4);7*1H2;/q;;;;;;;;+2/p-2

InChI key

RZLVQBNCHSJZPX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Zinc sulfate is a mineral salt broadly used as supplement for nutrition.

Packaging

1282100003 (3X1 kg); 1282101001 (1 kg); 1282105001 (5 kg); 1282108025 (25 kg)
1282100003 (Sample kit); 1282101001 (Plastic bottle); 1282105001 (Plastic bottle); 1282108025 (PE bag in plastic bucket)

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service