Skip to Content
Merck
All Photos(1)

Key Documents

128125

Sigma-Aldrich

Aloisine A

A cell-permeable pyrrolo-pyrazine compound that exerts anti-proliferative effects.

Synonym(s):

Aloisine A, RP107, 7- n-Butyl-6-(4-hydroxyphenyl)[5H]pyrrolo[2,3-b]pyrazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17N3O
CAS Number:
Molecular Weight:
267.33
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[nH]1c2nccnc2c(c1c3ccc(cc3)O)CCCC

InChI

1S/C16H17N3O/c1-2-3-4-13-14(11-5-7-12(20)8-6-11)19-16-15(13)17-9-10-18-16/h5-10,20H,2-4H2,1H3,(H,18,19)

InChI key

PRIGRJPRGZCFAS-UHFFFAOYSA-N

General description

A cell-permeable pyrrolo-pyrazine compound that exerts anti-proliferative effects. Acts as a potent, selective, reversible, and ATP-competitive inhibitor of cyclin-dependent kinases (Cdks; IC50 = 150 nM, 120 nM, 400 nM, and 200 nM for Cdk1/cyclin B, Cdk2/cyclin A, Cdk2/cyclin E, and Cdk5/p25, respectively), glycogen synthase kinase-3 (GSK-3; IC50 = 500 nM and 1.5 µM for GSK-3α, GSK-3β, respectively), and c-Jun N-terminal kinase (JNK; IC50 = ~ 3 - 10 µM). It inhibits several other enzymes (CK1, CK2, MAPKK, PKA, PKG, PKCs, and c-raf) poorly (IC50 ≥ 100 µM). Shown to arrest cells in both G1 and G2 phases (IC50 = 7 µM and 10.5 µM for undifferentiated human teratocarcinoma cells NT2 and differentiated postmitotic neurons hNT, respectively). Shown to selectively stimulate CFTR-dependent iodide efflux in wtCFTR-CHO, Calu-3 and F508del-CFTR-CF15 cells in the presence of 1 µM Forskolin (Cat. No. 344270) with high affinity (EC50 = 150 nM, 140 nM and 111 nM, respectively).
A cell-permeable, pyrrolo-pyrazine compound that exerts anti-proliferative effects. Acts as a potent, selective, reversible, and ATP-competitive inhibitor of cyclin-dependent kinases (Cdks; IC50 = 150 nM for Cdk1/cyclin B, 120 nM for Cdk2/cyclin A, 400 nM for Cdk2/cyclin E, and 160 nM for Cdk5/p25), glycogen synthase kinase-3 (GSK-3; IC50 = 500 nM for GSK-3α and 1.5 µM for GSK-3β), and c-Jun N-terminal kinase (JNK; IC50 ~3-10 µM). Also inhibits extracellular signal-regulated kinases (ERKs; IC50 = 18 µM for ERK1 and 22 µM for ERK2), PIM1 (IC50 >10 µM), and insulin receptor tyrosine kinase (IC50 = 60 µM) at higher concentrations. Does not significantly inhibit Cdk4/cyclin D1, CK1, CK2, MAPKK, PKA, PKG, PKCs, or c-raf (IC50 = ≥ 100 µM). Shown to arrest undifferentiated human teratocarcinoma cells (NT2) and differentiated post-mitotic neurons (hNT) in both G1 and G2 phases (IC50 = 7 µM and 10.5 µM, respectively). Also shown to selectively stimulate CFTR-dependent iodide efflux in wtCFTR-CHO, Calu-3 and F508del-CFTR-CF15 cells in the presence of 1 µM of Forskolin (Cat. No. 344270) with high affinity (EC50 = 150 nM, 140 nM and 111 nM, respectively).

Biochem/physiol Actions

Cell permeable: yes
EC50 = 150 nM, 140 nM and 111 nM, in stimulating CFTR-dependent iodide efflux in wtCFTR-CHO, Calu-3 and F508del-CFTR-CF15 cells in the presence of 1 µM Forskolin, respectively
Primary Target
Cyclin-dependent kinases
Product competes with ATP.
Reversible: yes
Target IC50: 150 nM, 120 nM, 400 nM, and 200 nM for Cdk1/cyclin B, Cdk2/cyclin A, Cdk2/cyclin E, and Cdk5/p25, respectively; 500 nM and 1.5 µM for GSK-3α, GSK-3β, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Noel, S., et al. 2006. J. Pharm. Exp. Ther. , 319, 349.
Mettey, Y., et al. 2003. J. Med. Chem.46, 222.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service