Skip to Content
Merck
All Photos(1)

Documents

126871

Sigma-Aldrich

Alsterpaullone, 2-Cyanoethyl - CAS 852529-97-0 - Calbiochem

A cell-permeable and reversible Alsterpaullone derivative that acts as a highly potent, ATP-competitive, selective inhibitor of Cdk1/cyclin B and GSK-3β.

Synonym(s):

Alsterpaullone, 2-Cyanoethyl - CAS 852529-97-0 - Calbiochem, 3-(6-Oxo-9-nitro-5,6,7,12-tetrahydroindolo[3,2-d][1]benzazepin-2-yl)propionitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H14N4O3
CAS Number:
Molecular Weight:
346.34
UNSPSC Code:
12352200

Assay

≥90% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light orange

solubility

DMSO: 10 mg/mL

shipped in

wet ice

storage temp.

2-8°C

General description

A cell-permeable and reversible Alsterpaullone (Cat. No. 126870) derivative that acts as a highly potent, ATP-competitive, selective inhibitor of Cdk1/cyclin B and GSK-3β (IC50 = 230 pM and 800 pM, respectively; [ATP] = 15 µM). Displays ~37-fold greater selectivity for Cdk1/cyclin B over Cdk5/p25 (IC50 = 30 nM). Shown to inhibit other kinases in a commercially available testing screen panel (pIC50 = −logIC50[M]; pIC50 = ~ 7.5, 7.0, 7.0, 6.5, 6.5, 6.0, and 6.0 for Cdk2/A, Cdk4/D1, GSK-3β, PDGFRβ, Src, VEGFR-2, and VEGFR-3, respectively; [ATP] = 1 µM).
A cell-permeable and reversible Alsterpaullone (Cat. No. 126870) derivative that acts as a highly potent and ATP-competitiveinhibitor of Cdk/GSK-3 family serine/threonine kinases (IC50 = 230 pM and 800 pM for cdk1/cyclin B and Gsk-3β, respectively; [ATP] = 15 µM). Displays ~130-fold greater selectivity for Cdk1/cyclin B over Cdk5/p25 (IC50 = 30 nM). Shown to inhibit tyrosine kinases only at higher concentrations (pIC50 = −logIC50[M]; pIC50 ~7.5, 7.0, 7.0, 6.5, 6.5, 6.0, and 6.0 for Cdk2/A, Cdk4/D1, GSK-3β, PDGFRβ, Src, VEGFR-2, and VEGFR-3, respectively; [ATP] = 1 µM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Cdkl/Gsk-3β, Cyclin B
Product competes with ATP.
Reversible: yes
Target IC50: 230 pM and 800 pM, [ATP] = 15 µM, against Cdk1/cyclin B and GSK-3β, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Preparation Note

Slight warming may be required for complete solubilization.

Reconstitution

Following reconstitution, aliquot and refrigerate (4°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Kunick, C., et al. 2005. ChemBioChem.6, 541.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service