Skip to Content
Merck
All Photos(1)

Documents

126850

Sigma-Aldrich

α1-PDX, Human, Recombinant, E. coli

Recombinant protein derived from the bioengineered human α1-antitrypsin gene fused to a His•Tag sequence and a FLAG-tag.

Synonym(s):

α₁-Antitrypsin Portland

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥80% (SDS-PAGE)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

solubility

water: 2.5 mg/mL

shipped in

wet ice

storage temp.

2-8°C

General description

Recombinant protein derived from the bioengineered human α1-antitrypsin gene fused to a His·Tag and a FLAG·Tag. Contains a minimal furin consensus sequence, RXXR, that effectively blocks the furin-dependent processing of protein precursors (Ki = 600 pM). α1-PDX can be used in extracellular applications to block the processing of furin substrates. α1-PDX has been shown to inhibit the processing of biologically significant proteins including viral envelope glycoproteins, bacterial toxins, receptors, and growth factors.
Recombinant protein derived from the bioengineered human α1-antitrypsin gene fused to a His•Tag sequence and a FLAG-tag. Contains a minimal furin consensus sequence, RXXR, that effectively blocks the furin-dependent processing of protein precursors (Ki = 600 pM). Extracellular application of α1-PDX can be used to inhibit the processing of biologically significant proteins including viral envelope glycoproteins, bacterial toxins, receptors, and growth factors. Useful for in vivo and in vitro furin inhibition assays.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Furin-dependent processing of protein precursors
Product does not compete with ATP.
Reversible: no
Target Ki: 600 pM against furin-dependent processing of protein precursors

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from PBS.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Bassi, D.E., et al. 2001. Proc. Natl. Acad. Sci. USA98, 10326.
Jean, F., et al. 2000. Proc. Natl. Acad. Sci. USA97, 2864.
Jean, F., et al. 1998. Proc. Natl. Acad. Sci. USA95, 7293.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service