Skip to Content
Merck
All Photos(2)

Documents

1.25172

Millipore

Eshmuno® CP-FT Resin

RoboColumn Eshmuno® CP-FT, volume 0.6 mL, prepacked column

Synonym(s):

RoboColumn Eshmuno® CP-FT, 0.6ml

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

ligand

(Sulfoisobutyl)

Quality Level

material

polypropylene housing

product line

Eshmuno®

form

prepacked column

parameter

≤8 bar pressure
400 cm/hr max. flow rate

column I.D. × L

5 mm × 30 mm

volume

0.6 mL

matrix active group

Polyvinyl Ether

mean particle size

50 μm

capacity

~70 mL, resin binding capacity

application(s)

mAb

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

General description

5 x 30mm

Features and Benefits

Eshmuno® CP-FT chromatography resin enables:

  • Superior flow-through removal of mAb aggregates
  • High product recoveries at high mass loadings
  • Significant reduction in resin and buffer volume
  • Smaller manufacturing footprint (smaller columns, buffer tanks, etc.)
  • Low salt process conditions eliminate the need for dilution before subsequent ion exchange steps
  • Significant reduction in processing volumes improves virus filtration and ultrafiltration processing economics
  • Rigid base bead enables higher flow rates and easier column packing
  • Strong chemical stability
  • Compatibility with 1 % (v/v) aqueous benzyl alcohol containing 150 mM NaCl and storage solutions 0.1 M NaOH

Packaging

Plastic bag(s)

Legal Information

Eshmuno is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service