Skip to Content
Merck
All Photos(3)

Key Documents

1.25065

Millipore

Eshmuno® Q Resin

MiniChrom Column Eshmuno® Q, volume 1 mL, prepacked column

Synonym(s):

MiniChrom Column Eshmuno® Q, 1 ml

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
Pricing and availability is not currently available.

ligand

(Trimethylammoniumethyl)

material

polypropylene housing

product line

Eshmuno®

form

prepacked column

parameter

≤8 bar pressure
1000 cm/hr max. flow rate

column I.D. × L

8 mm × 20 mm

volume

1 mL

matrix active group

Polyvinyl Ether

mean particle size

85 μm

capacity

~150 mL, resin binding capacity

application(s)

mAb
recombinant protein
vaccine development

separation technique

anion exchange

storage temp.

15-25°C

General description

8 x 20mm

Features and Benefits

Eshmuno® Q chromatography resin enables:
  • Purification of acidic and neutral proteins and peptides from different sources, including plasma

Superior productivity for downstream processing of biomolecules
  • High flow rate versus pressure-flow behavior
  • Excellent removal of impurities, such as HCP, leached Protein A, and viruses
  • Robust and safe packing procedures
  • Strong chemical stability

Analysis Note

Appearance: passes test

Legal Information

Eshmuno is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Learn more on mAb downstream processing, more specifically polishing chromatography and the relevant associated products.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service