Skip to Content
Merck
All Photos(1)

Documents

1.24839

Millipore

Pefabloc® SC (4-(2-Aminoethyl)-benzolsulfonylfluorid-hydrochloride)

(AEBSF) for biochemistry

Synonym(s):

Pefabloc® SC (4-(2-Aminoethyl)-benzolsulfonylfluorid-hydrochloride), (4-(2-aminoetyl)-benzenesulfonyl fluoride hydrochloride)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11ClFNO2S
CAS Number:
Molecular Weight:
239.69
MDL number:
UNSPSC Code:
12352107
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

solid

mp

175-177 °C

solubility

10 g/L

bulk density

200 kg/m3

storage temp.

2-8°C

InChI

1S/C8H10FNO2S.ClH/c9-13(11,12)8-3-1-7(2-4-8)5-6-10;/h1-4H,5-6,10H2;1H

InChI key

WRDABNWSWOHGMS-UHFFFAOYSA-N

Analysis Note

Assay (HPLC): ≥ 98 %
Identity (IR-spectrum): passes test

Legal Information

Pefabloc is a registered trademark of Pentapharm

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service