1.24644
Dihydronicotinamide adenine dinucleotide disodium salt
for biochemistry NADH-Na₂
Synonym(s):
Dihydronicotinamide adenine dinucleotide disodium salt, NADH-Na2, Dihydrodiphosphopyridine nucleotide disodium salt, Nicotinamide adenine dinucleotide reduced disodium salt (NADH2)
About This Item
Recommended Products
Quality Level
Assay
≥97.0% dry basis (enzymatic)
storage temp.
2-8°C
SMILES string
[Na+].[Na+].[P](=O)([O-])(O[P](=O)(OC[C@H]4O[C@H]([C@@H]([C@@H]4O)O)[n+]5cc(ccc5)\C(=N\[H])\[O-])O)OC[C@H]1O[C@H]([C@@H]([C@@H]1O)O)[n]2c3ncnc(c3nc2)N
InChI
1S/C21H27N7O14P2.2Na/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(32)14(30)11(41-21)6-39-44(36,37)42-43(34,35)38-5-10-13(29)15(31)20(40-10)27-3-1-2-9(4-27)18(23)33;;/h1-4,7-8,10-11,13-16,20-21,29-32H,5-6H2,(H5-,22,23,24,25,33,34,35,36,37);;/q;2*+1/p-1/t10-,11-,13-,14-,15-,16-,20-,21-;;/m1../s1
InChI key
GUIOTHWMFMEYPG-WUEGHLCSSA-M
Related Categories
General description
Analysis Note
Identity (IR-spectrum): passes test
Water (according to Karl Fischer): ≤ 6
Spec. Absorptivity A 1%/1cm (260 nm; 0.01 g/l; buffer pH 7.0; calculated on solvent-free substance): 200 - 215
Ethanol (GC): ≤ 6
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service