Skip to Content
Merck
All Photos(1)

Key Documents

12-241

Sigma-Aldrich

Phospho-Glycogen Synthase Peptide-2

lyophilized, for Kinase Assays

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41
Pricing and availability is not currently available.

Product Name

Phospho-Glycogen Synthase Peptide-2, Phospho-Glycogen Synthase Peptide-2 primarily used in Kinase Assays.

Quality Level

form

lyophilized

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

shipped in

wet ice

Quality

Routinely evaluated as a substrate of active GSK3β (Catalog # 14-306) to catalyze the incorporation of phosphate into phospho-glycogen synthase peptide-2.

Storage and Stability

Lyophilized: Stable for 2 years at -20°C from date of shipment. Rehydrated: Stable for 6 months at -20°C. Aliquot rehydrated solution to avoid repeated freezing and thawing.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Use of peptide substrates for affinity purification of protein-serine kinases.
Woodgett, J R
Analytical biochemistry, 180, 237-241 (1989)
Regulation of glycogen synthase kinase-3 in human skeletal muscle: effects of food intake and bicycle exercise
Wojtaszewski, J. F., et al
Diabetes, 50, 265-269 (2001)
C M Moxham et al.
The Journal of biological chemistry, 271(48), 30765-30773 (1996-11-29)
Mitogen-activated protein kinases (MAPKs) represent a conserved family of Ser/Thr protein kinases with central roles in intracellular signaling. Activation of three prominent members of the MAPK family, i.e. extracellular response kinases (ERK), jun N-terminal kinase (JNK), and p38, was defined
M Pap et al.
The Journal of biological chemistry, 273(32), 19929-19932 (1998-08-01)
Growth factor-dependent survival of a variety of mammalian cells is dependent on the activation of phosphatidylinositol (PI) 3-kinase and its downstream effector, the protein kinase Akt. Glycogen synthase kinase-3 (GSK-3) has been previously identified as a physiological target of Akt
S R Kimball et al.
The Journal of biological chemistry, 273(47), 30945-30953 (1998-11-13)
The present study was designed to investigate the mechanism through which leucine and histidine regulate translation initiation in L6 myoblasts. The results show that both amino acids stimulate initiation and coordinately regulate the activity of eukaryotic initiation factor eIF2B. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service