Skip to Content
Merck
All Photos(1)

Key Documents

1.20597

Millipore

CSB (Cronobacter Selective Broth)

according to ISO 22964, GranuCult®

Synonym(s):

Cronobacter Selective Broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74
Pricing and availability is not currently available.

Agency

according to ISO 22964

Quality Level

form

medium granules

packaging

pkg of 500 g

manufacturer/tradename

GranuCult®

storage condition

(Tightly closed. Dry)

technique(s)

microbiological culture: suitable

color

light yellow to gray

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

Cronobacter spp. (spp.)

General description

Cronobacter Selective Broth (CSB) contains high nutrient content with an enzymatic digest of animal tissue and meat extract creating good growth conditions for Cronobacter. Vancomycin inhibits the growth of Gram-positive flora, and sodium chloride maintains the osmotic balance of the medium. Upon fermentation of sucrose, Cronobacter produces a color change of the bromocresol indicator. However, this reaction is not always positive for Cronobacter in the given incubation time. Hence, it is necessary that every selective enrichment in CSB must be sub-cultured on Chromogenic Cronobacter Isolation (CCI) agar regardless of its color change.

Application

Cronobacter Selective Broth is suitable for the selective enrichment of Cronobacter spp. from food products and ingredients intended for human consumption and the feeding of animals and environmental samples in the area of food.

Features and Benefits

  • Safe and sustainable due to reduced risks associated with fine dust and toxic substance inhalation, resulting in a safer work environment.
  • Excellent wettability, solubility, and free flowing properties
  • Convenient, with minimal component separation and clumping, even under warm or humid conditions
  • High batch-to-batch reproducibility
  • Prolonged shelf life of up to five years
  • High number of test strains exceeding all regulatory demands
  • Granulation technology allowing many supplements to be included, with no need to add these separately

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Interview: Accurate microbiology QC methods for infant formula testing

Protocols

The new EN ISO 22964:2017 standard specifies a horizontal method for the detection of Cronobacter spp. in food, animal feed and environmental samples.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service