Skip to Content
Merck

Skip To

1.18348

Kjeldahl tablets

weight 3.5 g (tablet)without addition of Se and Hg

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NB.21
UNSPSC Code:
12352402
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

tablet

Quality Segment

specific analyte(s)

nitrogen

technique(s)

photometry: suitable

weight

3.5 g (tablet)without addition of Se and Hg

storage temp.

no temp limit

General description

(without addition of Se and Hg) 3.5 g/tablet

Application


  • Health risk assessment of arsenic in Realgar and NiuHuangJieDu Tablets based on pharmacokinetic study: This research utilizes Kjeldahl tablets to determine the nitrogen content in pharmaceutical compounds, specifically assessing the health risks associated with arsenic levels in traditional Chinese medicine. This study underlines the critical application of Kjeldahl analysis in ensuring the safety and efficacy of pharmaceutical products (Wu et al., 2018).

  • Application of infrared spectroscopy technique to protein content fast measurement in milk powder based on support vector machines: This study illustrates the use of Kjeldahl tablets for rapid protein quantification in milk powder, employing infrared spectroscopy enhanced by machine learning algorithms. The research highlights the effectiveness of Kjeldahl nitrogen analysis in food science, ensuring quality control and nutritional assessment in dairy products (Wu et al., 2008).


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
117958115348118469
description

weight 3.5 g (tablet)without addition of Se and Hg

description

-

description

weight 5 g (tablet)without addition of Se and Hg

description

(Missouri catalyst) 3.5g/tablet

technique(s)

photometry: suitable

technique(s)

photometry: suitable

technique(s)

photometry: suitable

technique(s)

photometry: suitable

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

form

tablet

form

tablet

form

tablet

form

tablet

storage temp.

no temp limit

storage temp.

-

storage temp.

no temp limit

storage temp.

no temp limit

specific analyte(s)

nitrogen

specific analyte(s)

nitrogen

specific analyte(s)

nitrogen

specific analyte(s)

nitrogen

weight

3.5 g (tablet)without addition of Se and Hg

weight

-

weight

5 g (tablet)without addition of Se and Hg

weight

-


Still not finding the right product?

Explore all of our products under Kjeldahl tablets


hcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters