1.16992
Potassium Test Strips, reflectometric
0.25-1.2 g/L (K), REFLECTOQUANT®
Synonym(s):
Potassium chemical test strips, Potassium test strip for instrument reader
About This Item
Recommended Products
product name
Potassium Test, reflectometric, 0.25-1.2 g/L (K), for use with REFLECTOQUANT®
description
quantitative
Quality Level
specific analyte(s)
potassium
measuring range
0.25-1.2 g/L (K)
compatibility
RQflex® 10 Plus
RQflex® 10
RQflex® 20
for use with REFLECTOQUANT®
detection method
reflectometric
storage temp.
15-25°C
General description
Analysis Note
Assay
- standard 1:
0.35 - 0.45 g/l
- standard 2: 0.62 - 0.78 g/l
- standard 3: 0.90 - 1.10 g/l
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 4 Oral
Storage Class Code
6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service