Skip to Content
Merck
All Photos(1)

Documents

1.16727

Supelco

Empty cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NB.35

compatibility

for use with RQflex® 10 Plus

detection method

reflectometric

General description

We offer you useful accessories. The empty cells can be used for the RQflex® 10 plus reflectometer. The package includes 100 disposable cells.

Legal Information

RQFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service