Skip to Content
Merck

Skip To

1.16141

Sucrose (Saccharose) Test Strips, reflectometric

0.25-2.50 g/L (sucrose), REFLECTOQUANT®, for use with RQflex® 20

Synonym(s):

Sucrose (Saccharose) chemical test strips, Sucrose (Saccharose) test strip for instrument reader

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Sucrose (Saccharose) Test, reflectometric, 0.25-2.50 g/L (sucrose), for use with REFLECTOQUANT®

description

quantitative

Quality Segment

analyte chemical class(es)

sugars

reaction suitability

reaction type: enzymatic reaction

measuring range

0.25-2.50 g/L (sucrose)

application(s)

beverage testing
food testing
milk and dairy products

compatibility

RQflex® 20, for use with REFLECTOQUANT®

detection method

reflectometric

storage temp.

2-8°C

General description

Method: reflectometric with test strips and reagent 0.25 - 2.50 g/l Reflectoquant®
The Reflectoquant Sucrose test is suitable for determination of sucrose in beverages, e. g. fruit/vegetable juice and refreshment drinks after appropriate sample pretreatment or dilution. Sucrose (saccharose) is cleaved by sucrose phosphorylase into fructose and glucose-1-phosphate. The latter is converted by phosphoglucomutase into glucose-6-phosphate. This is in turn oxidized by NAD under the catalytic effect of glucose-6-phosphate dehydrogenase to gluconate-6-phosphate. In the presence of diaphorase the NADH formed in the process reduces a tetrazolium salt to a blue formazan that is determined reflectometrically on the RQflex® 20. For the determination of only glucose please refer to our MQuant or Reflectoquant Glucose tests.

Application

Sucrose (Saccharose) Test Strips, reflectometric can be used:      
</UL>
In the beverage industry, particularly for determining sucrose levels in soft drinks and fruit juices. This helps manufacturers ensure product consistency and compliance with

Analysis Note

function control: entspricht
Assay
- standard 1:
0.26 - 0.34 mg/l
- standard 2: 1.12 - 1.38 mg/l
- standard 3: 1.80 - 2.20 mg/l

Other Notes

Replaces: 1161410007; 1.16141.0007

Legal Information

REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
RQFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.169871.167201.16127
detection method

reflectometric

detection method

reflectometric

detection method

reflectometric

detection method

reflectometric

compatibility

RQflex® 20, for use with REFLECTOQUANT®

compatibility

RQflex® 20, for use with REFLECTOQUANT®

compatibility

RQflex® 20, for use with REFLECTOQUANT®

compatibility

RQflex® 20, for use with REFLECTOQUANT®

reaction suitability

reaction type: enzymatic reaction

reaction suitability

-

reaction suitability

-

reaction suitability

reaction type: enzymatic reaction

application(s)

beverage testing
food testing
milk and dairy products

application(s)

boiler and cooling water
food testing
wastewater

application(s)

beverage testing
food testing
milk and dairy products

application(s)

beverage testing
food testing
milk and dairy products

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

description

quantitative

description

quantitative

description

quantitative

description

quantitative


Storage Class

10-13 - German Storage Class 10 to 13



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters