Skip to Content
Merck

Skip To

1.16124

Magnesium Test Strips, reflectometric

5-100 mg/L (Mg), REFLECTOQUANT®, for use with RQflex® 20

Synonym(s):

Magnesium chemical test strips, Magnesium test strip for instrument reader

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Magnesium Test, reflectometric, 5-100 mg/L (Mg), for use with REFLECTOQUANT®

description

quantitative

Quality Segment

specific analyte(s)

magnesium

measuring range

5-100 mg/L (Mg)

application(s)

agriculture
boiler and cooling water
drinking water
food testing
ground and surface water
mineral water

compatibility

RQflex® 20, for use with REFLECTOQUANT®

detection method

reflectometric

storage temp.

2-8°C

General description

Method: reflectometric with test strips 5 - 100 mg/l Mg Reflectoquant®
The Reflectoquant Magnesium test is suitable for determining magnesium ions in groundwater and surface water, drinking water, mineral water, boiler water and food after pretreatment. Magnesium ions react with phthalein purple to form a violet dye that is determined reflectometrically on the RQflex® 20.

Analysis Note

function control: conforms
Assay
- standard 1:
8 - 12 mg/l
- standard 2: 36 - 44 mg/l
- standard 3: 72 - 88 mg/l

Legal Information

REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
RQFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.169821.167201.16995
specific analyte(s)

magnesium

specific analyte(s)

iron

specific analyte(s)

-

specific analyte(s)

nitrate

detection method

reflectometric

detection method

reflectometric

detection method

reflectometric

detection method

reflectometric

compatibility

RQflex® 20, for use with REFLECTOQUANT®

compatibility

RQflex® 20, for use with REFLECTOQUANT®

compatibility

RQflex® 20, for use with REFLECTOQUANT®

compatibility

RQflex® 20, for use with REFLECTOQUANT®

storage temp.

2-8°C

storage temp.

15-25°C

storage temp.

2-8°C

storage temp.

2-8°C

measuring range

5-100 mg/L (Mg)

measuring range

0.5-20 mg/L (Fe2+)

measuring range

1-100 mg/L (glucose)

measuring range

3-90 mg/L (NO3-)

description

quantitative

description

quantitative

description

quantitative

description

quantitative


Storage Class

11 - Combustible Solids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters