1.15937
Fuchsin (C.I. 42510)
for microscopy Certistain®
Synonym(s):
Fuchsin (C.I. 42510)
About This Item
Recommended Products
Quality Level
form
solid
composition
Dye, ≥80% spectrophotometric assay
pH
5-6 (25 °C, 1 g/L in H2O)
mp
235 °C (decomposition)
solubility
4 g/L
bulk density
500 kg/m3
εmax
2150-2710 at 549-552 nm in 50% ethanol at 0.003 g/L
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
2-30°C
InChI
1S/C20H19N3.ClH/c1-13-12-16(6-11-19(13)23)20(14-2-7-17(21)8-3-14)15-4-9-18(22)10-5-15;/h2-12,23H,21-22H2,1H3;1H
InChI key
AXDJCCTWPBKUKL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
The cell wall of acid-fast bacteria has a high proportion of wax and lipids and hence absorbs dyes only very slowly. As a measure to enhance the absorption of the fuchsin dye and thus the formation of the mycolate-fuchsin complex in the cell wall, the carbolfuchsin solution applied to the specimen is normally heated to evaporation point. If the Kinyoun carbolfuchsin solution is used, the heating of the solution is dispensable. Consequently, the release of hazardous phenolic vapors will be avoided.
Once the acid-fast bacteria have absorbed the fuchsin dye, it is virtually impossible to decolorize them again, even when they are intensively treated with a decolorizing solution such as e. g. hydrochloric acid in ethanol. Accordingly, acid-fast bacteria are termed as acid- and alcohol-fast for staining, and are stained red in the microscopic visualization. Correspondingly, all non-acid-fast microorganisms are counterstained with an appropriate dye. In the present operating instructions is counterstained accordance with methylene blue.
Pretreatment of the specimens with Sputofluol™ dissolves the bacteria from the surrounding viscid sputum and cell material. This product is registered as IVD and CE marked. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.
Analysis Note
Identity (UV/VIS-Spectrum): passes test
Absorption maximum λ max (ethanol 50 %): 549 - 552 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.003 g/l; ethanol 50 %): 2150 - 2710
TLC-Test: passes test
Loss on drying (110 °C): ≤ 15 %
Suitability for microscopy: passes test
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service