Skip to Content
Merck
All Photos(1)

Key Documents

1.14902

Supelco

Flat-bottomed tubes with screw caps

for use with MQuant® titrimetric and colormetric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24
Pricing and availability is not currently available.

Product Name

Flat-bottomed tubes with screw caps, for use with MQUANT® titrimetric and colormetric tests

compatibility

for use with MQUANT® titrimetric and colormetric tests

detection method

colorimetric

storage temp.

15-25°C

General description

We offers you useful accessories. These short flat-bottomed tubes can be used for our colorimetric and titrimetric MQuant® test kits with color card comparator (formally MColortest®). The screw caps are included.
for titrimetric and colorimetric MQuant® tests

Analysis Note

Application test: conforms

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

BCA protein assay or Smith assay to determine protein concentration in a sample using Spectroquant® Prove photometers.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service