Skip to Content
Merck

Skip To

1.14792

Silicate (Silicic Acid) Test Kit, colorimetric

0.3-10 mg/L (Si), MQuant®

Synonym(s):

Silicate Visual Test Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
41116116
NACRES:
NB.21
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Silicate (Silicic Acid) Test, colorimetric, 0.3-10 mg/L (Si), for use with MQUANT®

specific analyte(s)

silicate

Quality Level

measuring range

0.3-10 mg/L (Si)

compatibility

for use with MQUANT®

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric with color-disk comparator 0.3 - 0.6 - 1.0 - 1.5 - 2 - 3 - 5 - 7 - 10 mg/l Si MQuant®
The MQuant® Silicate (Silicic Acid) test (formally MColortest®) can be used for the determination of silicate e.g. in:
  • Groundwater, surface water, and seawater
  • Drinking water and mineral water
  • Industrial and process water
  • Boiler water and boiler feed water
  • Wastewater and percolating water

Method
In sulfuric solution silicate ions react with molybdate ions to form a yellow heteropoly acid. This is reduced to silicomolybdenum blue. The silicate concentration is measured semiquantitatively by visual comparison of the color of the measurement solution with the color fields of a color disk. The test evaluates the color reaction based on the transmitted light method. So even turbid and colored water samples can be analyzed without further preparation.
The color disk is made of very durable, lightfast plastic, thus suitable for industrial areas and wet environments.
Using the refill pack 1.18323 containing all the reagents, the original test kit can be used for a very long time, making it a particularly economical and ecological system.

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.144101.018131.14794
specific analyte(s)

silicate

specific analyte(s)

silicate

specific analyte(s)

silicate

specific analyte(s)

silicate

detection method

colorimetric

detection method

colorimetric

detection method

photometric (Silicomolybdenum blue)

detection method

photometric (Silicomolybdenum blue)

measuring range

0.3-10 mg/L (Si)

measuring range

0.01-0.25 mg/L (Si), 0.02-0.53 mg/L (SiO2)

measuring range

0.12-116.85 μg/L (Si), 0.2-233.7 μg/L (Si), 0.25-250.00 μg/L (SiO2), 0.5-500.0 μg/L (SiO2)

measuring range

0.005-5.00 mg/L (Si), 0.011-10.70 mg/L (SiO2)

storage temp.

15-25°C

storage temp.

15-25°C

storage temp.

15-25°C

storage temp.

15-25°C

compatibility

for use with MQUANT®

compatibility

for use with MCOLORTEST®

compatibility

for use with Spectroquant® Move 100, for use with Spectroquant® Nova 60 A, for use with Spectroquant® Prove 100 (plus), for use with Spectroquant® Prove 300 (plus), for use with Spectroquant® Prove 600 (plus)

compatibility

for use with Spectroquant® Move 100, for use with Spectroquant® Nova 60 A, for use with Spectroquant® Prove 100 (plus), for use with Spectroquant® Prove 300 (plus), for use with Spectroquant® Prove 600 (plus)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100


Still not finding the right product?

Explore all of our products under Silicate (Silicic Acid) Test


signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A - Skin Sens. 1

supp_hazards

Storage Class

8B - Non-combustible, corrosive hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content





Global Trade Item Number

SKUGTIN
114792000104022536156743

Questions

Reviews

No rating value

Active Filters