Skip to Content
Merck
All Photos(1)

Documents

1.14582

Millipore

mEC-broth with Novobiocin

for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

Quality Level

form

medium granules (dehydrated (DCM))

manufacturer/tradename

GranuCult®

technique(s)

microbiological culture: suitable

pH

6.7-7.1 (25 °C, 36.7 g/L in H2O, after autoclaving)

solubility

36.7 g/L

application(s)

food and beverages
microbiology
pathogen testing

storage temp.

15-25°C

suitability

Escherichia coli

Analysis Note

Appearance (clearness): clear
Appearance (color): yellowish to yellowish-braun
pH-value (25 °C): 6.7 - 7.1
Inoculum on reference medium (Escherichia coli ATCC 35150 (O157)): ≤ 100
Inoculum on reference medium (Escherichia coli ATCC 11775 (WDCM 00090)): ≤ 100
Inoculum on reference medium (Enterococcus faecalis ATCC 19433 (WDCM 00009)): ≥ 10000
Inoculum on reference medium (Enterococcus faecalis ATCC 29212 (WDCM 00087)): ≥ 10000
Inoculum on reference medium (Proteus hauseri ATCC 13315): ≥ 10000
Growth (Escherichia coli ATCC 35150 (O157)): >10 transparent colonies with a pale yellowish-brown appearance on SMAC agar
Growth (Escherichia coli ATCC 11775 (WDCM 00090)): >10 pink colonies on SMAC Agar
Growth (Enterococcus faecalis ATCC 19433 (WDCM 00009)): total inhibition on TSA
Growth (Enterococcus faecalis ATCC 29212 (WDCM 00087)): total inhibition on TSA
Mixed inoculum of E.coli, P.hauseri and E.faecalis
Additionaly monoculture of E.faecalis
Confirmation on SMAC agar resp. Tryptic soy agar
Incubation:
mEC broth: 18 - 24 hours at 41,5 ± 1 °C aerobic
Confirmation on SMAC agar resp. Tryptic Soy Agar: 18 - 24 hours at 35 ± 1 °C aerobic

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service