1.14566
Zinc Cell Test
photometric, 0.20-5.00 mg/L (Zn), Spectroquant®
About This Item
Recommended Products
product line
Spectroquant®
Quality Level
usage
sufficient for 25 tests
specific analyte(s)
zinc
measuring range
0.20-5.00 mg/L (Zn)
technique(s)
photometry: suitable
compatibility
for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600
detection method
photometric (PAR)
storage temp.
15-25°C
General description
This Spectroquant®Zinc test allows the accurate quantification of zinc ions in various waters. In alkaline solution zinc ions react with pyridylazoresorcinol (PAR) to form a red complex that is determined photometrically.
MQuant® test strips can be used for pre-testing a sample in an economic and quick way to estimate the zinc concentration and a possible dilution factor prior to using a quantitative method such as the Spectroquant® test.
All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.
The Spectroquant®Cell Tests come with prefilled 16 mm round cells as well as all the required reagents to perform the analysis according to the procedure described in the accompanying instruction leaflet.
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Chronic 3 - Carc. 1B - Eye Dam. 1 - Muta. 2 - Skin Corr. 1 - Skin Sens. 1 - STOT SE 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
Photometric determination subsequent to acid mineralisation with sulfuric acid and Perhydrol
Photometric determination with pyridylazoresorcinol (PAR) subsequent to acid extraction
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service