Skip to Content
Merck
All Photos(2)

Key Documents

1.14226

Millipore

Anaeroclip®

For use in providing a mechanical closure of Anaerocult products

Synonym(s):

Closure clips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NB.14
Pricing and availability is not currently available.

Quality Level

feature

ready-to-use

packaging

pkg of 25 units

application(s)

food and beverages
microbiology

compatibility

for use with

  • Anaerotest (1.15112)
  • Anaerocult A mini (1.32369)
  • Anaerocult P (1.32382)

storage temp.

15-25°C

suitability

anaerobic bacteria

General description

Anaeroclip®, an accessory of the Anaerocult system, is a simple mechanical clip. The Anaeroclip® closure clips enable users to seal the bags without a special foil sealing instrument and can be used several times.

Application

Anaeroclip® is used to seal the Anaerocult A Mini, P, IS, and C Mini incubation bags.

Compatibility

Compatible with Anaerotest (1.15112), Anaerocult A mini (1.32369), and Anaerocult P (1.32382)

Legal Information

ANAEROCLIP is a registered trademark of Merck KGaA, Darmstadt, Germany
ANAEROCULT is a trademark of Merck KGaA, Darmstadt, Germany
ANAEROTEST is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Anaerocult® system supports reliable anaerobe cultivation for microbiological research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service