Skip to Content
Merck
All Photos(1)

Key Documents

1.12352

Sigma-Aldrich

Tantalum(V) chloride

99+

Synonym(s):

Tantalum(V) chloride, Tantalum pentachloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cl5Ta
CAS Number:
Molecular Weight:
358.21
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-755-6
grade:
for analytical purposes
Pricing and availability is not currently available.

grade

for analytical purposes

Quality Level

potency

1900 mg/kg LD50, oral (Rat)

bp

239 °C/1013 hPa

mp

216-217 °C

density

3.68 g/cm3 at 25 °C

bulk density

2400 kg/m3

storage temp.

no temp limit

InChI

1S/5ClH.Ta/h5*1H;/q;;;;;+5/p-5

InChI key

OEIMLTQPLAGXMX-UHFFFAOYSA-I

Analysis Note

Identity (elemental analysis): conforms
Assay (gravimetrically): ≥ 99.8 %

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service