Skip to Content
Merck
All Photos(1)

Documents

1.12349

Sigma-Aldrich

Tantalum powder

98+

Synonym(s):

Tantalum powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ta
CAS Number:
Molecular Weight:
180.95
MDL number:
UNSPSC Code:
12141741
EC Index Number:
231-135-5
NACRES:
NA.22

Quality Level

form

solid

autoignition temp.

>250 °C

potency

>2000 mg/kg LD50, oral (Rat)

pH

7 (20 °C in H2O, aqueous suspension)

mp

2996 °C

solubility

<0.010 g/L

density

16.6 g/cm3 at 20 °C

bulk density

500 kg/m3

storage temp.

no temp limit

InChI

1S/Ta

InChI key

GUVRBAGPIYLISA-UHFFFAOYSA-N

Related Categories

Analysis Note

Identity (elemental analysis): passes test
Assay: ≥ 98.0 %

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service