Skip to Content
Merck
All Photos(1)

Documents

1.10854

Supelco

Formic acid

90%, for determination of viscosity acc. to DIN EN ISO 307

Synonym(s):

Methanoic acid solution, Formylic acid solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352106
NACRES:
NA.21

product name

Formic acid 90 %, for determination of viscosity acc. to DIN EN ISO 307

Quality Level

Assay

90.00-90.15% (acidimetric)

form

liquid

autoignition temp.

485 °C

expl. lim.

10-45.5 % (v/v)

impurities

≤0.050% Acetic acid (CH3COOH)

bp

107 °C/1013 hPa

mp

-9 °C

transition temp

flash point 71 °C

density

1.2 g/cm3 at 20 °C

storage temp.

15-25°C

Application

  • Quantitative high-throughput determination of endogenous retinoids in human plasma using triple-stage liquid chromatography/tandem mass spectrometry.: This study by Gundersen et al. (2007) presents a method for the high-throughput quantitative determination of endogenous retinoids in human plasma. The research utilizes triple-stage liquid chromatography/tandem mass spectrometry, highlighting the significance of formic acid in enhancing the sensitivity and accuracy of retinoid measurement (Gundersen et al., 2007).

Analysis Note

Assay (acidimetric): 90.00 - 90.15 %
Acetic acid (CH₃COOH): ≤ 0.050 %

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

159.8 °F

Flash Point(C)

71 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service