1.10841
p-Toluidine
GR for analysis
Synonym(s):
4-Aminotoluene, 4-Methylaniline, 4-Aminotoluene, 4-Methylaniline
About This Item
200 °C (lit.)
Recommended Products
vapor density
3.9 (vs air)
Quality Level
Assay
≥99.0% (GC)
form
solid
autoignition temp.
899 °F
potency
336 mg/kg LD50, oral (Rat)
890 mg/kg LD50, skin (Rabbit)
expl. lim.
6.6 %
dilution
(for analytical testing)
bp
200 °C/1,013 hPa
200 °C (lit.)
mp
41-46 °C (lit.)
transition temp
flash point 87 °C
density
0.973 g/mL at 25 °C (lit.)
storage temp.
2-30°C
SMILES string
Cc1ccc(N)cc1
InChI
1S/C7H9N/c1-6-2-4-7(8)5-3-6/h2-5H,8H2,1H3
InChI key
RZXMPPFPUUCRFN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- Preparation of Porous Organic Cage and its use as Chiral Stationary Phase for Capillary Electrochromatography: Research on p-toluidine-derived chiral stationary phases for enhanced electrochromatographic separations, emphasizing its utility in chiral drug analysis and purification (Zheng et al., 2023).
- Comprehensive N- and O-Glycoproteomic Analysis of Multiple Chinese Hamster Ovary Host Cell Lines: Application of p-toluidine in the detailed glycoproteomic analysis, demonstrating its role in enhancing detection and analysis techniques in cellular biology (Wang et al., 2022).
- Degradation of Isoproturon in vitro by a Mix of Bacterial Strains Isolated from Arable Soil: Utilizing p-toluidine-related methodologies for environmental monitoring and degradation studies, showcasing its importance in agricultural chemistry (Duc, 2022).
Analysis Note
Identity (IR-spectrum): passes test
Melting range: 43 - 46
o-Toluidine (GC): ≤ 0.1 %
m-Toluidine (GC): ≤ 0.2 %
Performance test (for the determination of hydroxy methylfurfurale): passes test
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Skin Sens. 1A
Storage Class Code
6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
188.6 °F - closed cup
Flash Point(C)
87 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service