Skip to Content
Merck
All Photos(1)

Documents

1.10757

Millipore

Silica gel 60 F₂₅₄ (0.063-0.200 mm)

adjusted to 40% relative humidity, suitable for dry-column chromatography

Synonym(s):

Silica gel

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41115711

Quality Level

form

solid

pH

6.5-7.5 (20 °C, 100 g/L in H2O, slurry)

bp

2230 °C

mp

>1600 °C

bulk density

400‑500 kg/m3

storage temp.

no temp limit

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Silica gel 60 F254 (0.063-0.200 mm) provides users with:

  • High adsorptive capacity
  • High chemical inertness and non-toxicity.
  • Protective moisture-sensitive substances during storage and transport or for maintaining the dryness of anhydrous solvents.
  • Granulates in different sizes or beads

Packaging

Plastic bottle

Analysis Note

pH-value (10% suspension): 6.5 - 7.5
Fe (Iron): ≤ 0.02%
Loss on drying (150 °C): 8.8 - 9.4%
Flow time: <1 minute / 10 cm
Particle size: 90% between 0.063 - 0.200 mm
Fluorescence intensity: conforms

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service